Robotic tips for Tecan®
903-252, 200 µl, -, -, Black, 10 racks, 96 each
Katalogové číslo 736-0055
Dodavatel: Thermo Fisher Scientific
Robotic tips for Tecan®
Katalogové číslo 736-0055
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
High quality tips, especially intended for research in molecular biology laboratories.
- Transparent or black PP
- Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
- Certified free of RNase, DNase, DNA and pyrogens
- Sterile tips: Sterilised by gamma radiation
- Wide aperture tips: Designed for applications with more viscous samples
Ordering information: Available in boxes of 10 racks or 50 racks (Econopak).
Specifikace
- Balené:10 racks, 96 each
- Sterilní:-
- Objem:200 µl
- Druh špičky:903-252
- Barva špičky:Black
- Filtr:-
- Cat. No.:736-0055
- Certifikát ochrany životního prostředí:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact