Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
10, 200 ml, 14/15, 0,15 ± ml
  POULRB258/WAC/H/2
Volumetric flasks, class A
  POULRB258/WAC/H/2
Volumetric flasks, class A
Volumetric flasks, class A
10, 200 ml, 14/15, 0,15 ± ml
1 040,00 Kč
1 * 2 KS
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
  • NZ:
    14/15
  • Kapacita:
    200 ml
  • Sekvence:
    10
  • Tolerance:
    0,15 ± ml