Orders LinkContactUsLinkComponent
Burettes, removable stopcock
Burettes, removable stopcock
05, Glass jet for 25 and 50 ml burettes
Katalogové číslo BIBBBRB284
Dodavatel:   SciLabware
Burettes, removable stopcock
Katalogové číslo BIBBBRB284
Dodavatel:  SciLabware
Burettes, removable stopcock
Burettes, removable stopcock
05, Glass jet for 25 and 50 ml burettes
Přejít na:
Price
747,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
Soda lime glass, class B, removable PTFE stopcock.
  • Flared top for safer filling
  • Blue enamel graduations
BS 846
Certifications: BS 846
Specifikace
  • Popis:
    Glass jet for 25 and 50 ml burettes
  • Sekvence:
    05