Orders LinkContactUsLinkComponent
PYREX®/SCILABWARE Extraction Thimbles
PYREX®/SCILABWARE Extraction Thimbles
01, 20×95 mm, Porosity grade 1
Catalog # BIBB3840/02M
Supplier:   SciLabware
PYREX®/SCILABWARE Extraction Thimbles
Catalog # BIBB3840/02M
Supplier:  SciLabware
PYREX®/SCILABWARE Extraction Thimbles
PYREX®/SCILABWARE Extraction Thimbles
01, 20×95 mm, Porosity grade 1
Price
9 360,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Borosilicate glass.
Specifications
  • Vnitř. Ø×V:
    20×95 mm
  • Type:
    Porosity grade 1
  • Sekvence:
    01