Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
02, 24/29 NS, 19/26 NS
  201-3001
 :   SciLabware
Multiple adapters, Quickfit®
  201-3001
Multiple adapters, Quickfit®
02, 24/29 NS, 19/26 NS
1 680,00 Kč
1 * 1 KS
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two necks. One vertical and one angled at 45°.
  • Rozměry zábrusu:
    24/29 NS
  • Velikost objímky:
    19/26 NS
  • Sekvence:
    02