Multiple adapters, Quickfit®
01, 19/26 NS, 19/26 NS
Katalogové číslo 201-3000
Dodavatel: SciLabware
Přejít na:
Product Details & Documents
PYREX®, borosilicate glass
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With two necks. One vertical and one angled at 45°.
Specifikace
- Rozměry zábrusu:19/26 NS
- Velikost objímky:19/26 NS
- Sekvence:01