Swabs tubes
Plain swabs, Wood/cotton, +
GREISWA6_1000
Swab in tubes suitable for the collection and transport of non-human bacteriological, serological, and cytological samples.
- Sterilised by irradiation
- Swab tubes made of polystyrene
Useful for hygienic controls in veterinary research as well as in environmental sampling.
: Individually wrapped.
: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
- Sterilní:+
- Popis:Plain swabs
- Materiál:Wood/cotton