Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
10, 96-well microplate, U-bottom, MICROLON® 200, 40 - 280 µl, 323 µl
Katalogové číslo 738-0004
Dodavatel:   Greiner Bio-One
Microplates, 96-well
Katalogové číslo 738-0004
Dodavatel:  Greiner Bio-One
Microplates, 96-well
Microplates, 96-well
10, 96-well microplate, U-bottom, MICROLON® 200, 40 - 280 µl, 323 µl
Přejít na:
Price
2 280,00 Kč
Přihlásit se pro dostupnost
1 * 40 KS
Product Details & Documents
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
  • Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
  • Free of DNase, RNase and human DNA, and are non-pyrogenic
Specifikace
  • Popis:
    96-well microplate, U-bottom, MICROLON® 200
  • Sekvence:
    10
  • Pracovní objem:
    40 - 280 µl
  • Celkový objem:
    323 µl
  • Cat. No.:
    738-0004