Connectors
02, Fittings, external thread for pipes and valves, 3/4"
Katalogové číslo SEMA4367
Dodavatel: SEMADENI
Přejít na:
Product Details & Documents
Connectors for easy installation on any plastic containers by means of counter screw.
- Male thread for fittings or hose connection
- Good chemical resistance to acids and solvents
Specifikace
- Závit:3/4"
- Popis:Fittings, external thread for pipes and valves
- Sekvence:02