Orders LinkContactUsLinkComponent
Trypsin
Trypsin
10, Trypsin-EDTA 1X, with sodium chloride, lyophilised, 100 g
Katalogové číslo P0940-100GR
Dodavatel:   Biowest
Trypsin
Katalogové číslo P0940-100GR
Dodavatel:  Biowest
Číslo CAS:  
Trypsin
Trypsin
10, Trypsin-EDTA 1X, with sodium chloride, lyophilised, 100 g
Přejít na:
Číslo CAS:  
Price
4 730,00 Kč
Přihlásit se pro dostupnost
1 * 100 g
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifikace
Specifikace
  • Bal.:
    100 g
  • Popis:
    Trypsin-EDTA 1X, with sodium chloride, lyophilised
  • Sekvence:
    10