Orders LinkContactUsLinkComponent
Trypsin
Trypsin
08, Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red, 500 mL
Katalogové číslo L0940-500
Dodavatel:   Biowest
Trypsin
Katalogové číslo L0940-500
Dodavatel:  Biowest
Číslo CAS:  
Trypsin
Trypsin
08, Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red, 500 mL
Přejít na:
Číslo CAS:  
Price
1 350,00 Kč
Přihlásit se pro dostupnost
1 * 500 mL
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifikace
Specifikace
  • Bal.:
    500 mL
  • Popis:
    Trypsin-EDTA 1X in PBS, without calcium, without magnesium, without phenol red
  • Sekvence:
    08