Adapters, expansion
02, , Expanze, 14/23 NS, 29/32 NS, ,
Katalogové číslo WITG0.510.003
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Borosilicate glass 3.3
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapters.
Certifications: According to DIN 12257.
Specifikace
- Rozměry zábrusu:14/23 NS
- Velikost objímky:29/32 NS
- Type:Expanze
- Sekvence:02