Orders LinkContactUsLinkComponent
Containers with screw cap
Containers with screw cap
04, 200 ml, Labelled, Individually wrapped, PS
Catalog # ANICC200S220
Supplier:   Anicrin
Containers with screw cap
Catalog # ANICC200S220
Supplier:  Anicrin
Containers with screw cap
Containers with screw cap
04, 200 ml, Labelled, Individually wrapped, PS
Price
2 200,00 Kč
Log in for availability
1 * 150 KS
Product Details & Documents
Containers made of PS or PP with colour coded screw caps.
  • Good chemical resistance
  • Good temperature resistance
Ordering information: Specify the cap colour (red, yellow and white) required.
Caution: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Documents
Specifications
  • Balené:
    Individually wrapped
  • Verze:
    Labelled
  • Kapacita:
    200 ml
  • Materiál:
    PS
  • Sekvence:
    04