Microscope adapters for micromanipulators
, Zeiss Axiovert® 10/35
Katalogové číslo 631-0224
Dodavatel: EPPENDORF
Přejít na:
Product Details & Documents
Microscope adapters for InjectMan® NI 2 and TransferMan® NK 2. The universal stand is suitable for mounting manipulators on upright or stereo microscopes, independent of the microscope tripod used.
Specifikace
- Popis:Zeiss Axiovert® 10/35