Multiple or single cuvette segments
Cuvettes in bulk segments, PMMA
Katalogové číslo KART1501
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
These twelve position cuvettes segments have been specially designed for use with the Cobas Mira® analyser.
Packaging: In cases of bulk segments (450) or 30 pre-racked segments (15).
Specifikace
- Popis:Cuvettes in bulk segments
- Materiál:PMMA