Orders LinkContactUsLinkComponent
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
6000 ml, 29/32 NS, 29/32 NS
Catalog # WITG0.772.006
Supplier:   witeg Labortechnik
Flasks, round bottom, with three parallel necks
Catalog # WITG0.772.006
Supplier:  witeg Labortechnik
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
6000 ml, 29/32 NS, 29/32 NS
Price
9 610,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Specifications
  • Kapacita:
    6000 ml
  • Boční hrdla:
    29/32 NS
  • Centrální hrdlo:
    29/32 NS