HEPES
3368, Glass bottle for solids, 50 g
Katalogové číslo ICNA0219454950
Dodavatel: MP Biomedicals
Přejít na:
Product Details & Documents
Synonym: N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid) Cell Culture Reagent Crystalline Free Acid Zwitterionic Buffer useful in the pH range 6.8-8.2. pKa=7.55 at 25°C
pH: 4.0 - 5.5 (1% aq soln)
Synonyma: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
- Číslo CAS:7365-45-9
- Vzorec:C₈H₁₈N₂O₄S
- MDL číslo:MFCD00006158
- Molecular Weight:238.31
- Merck Index:13,04672
- Bod tání:210…215 °C
- Hustota:1,448 g/cm³ (20 °C)
- :Density: 1,448 g/cm³ (20 °C)
- Teplota pro skladování:Ambient
- EINECS:230-907-9
Specifikace
Specifikace
- Typ balení:Glass bottle for solids
- Bal.:50 g
- Sekvence:3368
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



