HEPES
3236, Glass bottle for solids, 10 g
ICNA0219454910
Synonym: N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid) Cell Culture Reagent Crystalline Free Acid Zwitterionic Buffer useful in the pH range 6.8-8.2. pKa=7.55 at 25°C
pH: 4.0 - 5.5 (1% aq soln)
: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
- :7365-45-9
- :C₈H₁₈N₂O₄S
- :MFCD00006158
- :238.31
- :13,04672
- :210…215 °C
- :1,448 g/cm³ (20 °C)
- :Density: 1,448 g/cm³ (20 °C)
- :Ambient
- :230-907-9
- Typ balení:Glass bottle for solids
- Bal.:10 g
- Sekvence:3236
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



