Orders LinkContactUsLinkComponent
Stages
04, Black and white stage plate, Ø: 95 mm
Catalog # 630-2779
Supplier:   MOTIC
Stages
Catalog # 630-2779
Supplier:  MOTIC
Stages
Stages
04, Black and white stage plate, Ø: 95 mm
Price
225,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    Black and white stage plate, Ø: 95 mm
  • Sekvence:
    04