pH indicator papers, MQuant®, Supelco®
Special indicator, 3,8 - 5,4, 0,2 - 0,3
: Merck
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
- pH stupnice:0,2 - 0,3
- pH rozsah:3,8 - 5,4
- Popis:Special indicator
- Cat. No.:1.09555.0003
- M-Clarity quality level:MQ100