pH indicator papers, Supelco®
Neutralit®, 5,5 - 9,0, 0,5
Dodavatel: Merck
Přejít na:
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
Documents
Specifikace
- pH stupnice:0,5
- pH rozsah:5,5 - 9,0
- Popis:Neutralit®
- Cat. No.:1.09564.0003
- M-Clarity quality level:MQ100