Orders LinkContactUsLinkComponent
Filter discs, DURAN®
Filter discs, DURAN®
40, 2, 30 mm, 3,4 mm
Katalogové číslo SCOT251530202
Dodavatel:   DWK Life Sciences
Filter discs, DURAN®
Katalogové číslo SCOT251530202
Dodavatel:  DWK Life Sciences
Filter discs, DURAN®
Filter discs, DURAN®
40, 2, 30 mm, 3,4 mm
Přejít na:
Price
1 670,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.
  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design
Made from clear DURAN® borosilicate glass 3,3.
Specifikace
  • Ø:
    30 mm
  • Porozita:
    2
  • Sekvence:
    40
  • Tloušťka:
    3,4 mm