Orders LinkContactUsLinkComponent
Hexagonal nuts with inner thread
Hexagonal nuts with inner thread
hexagonal nut with inner thread, for Ø 27 mm, ½", PE
Katalogové číslo BURK8537-0012
Dodavatel:   BURKLE
Hexagonal nuts with inner thread
Katalogové číslo BURK8537-0012
Dodavatel:  BURKLE
Hexagonal nuts with inner thread
Hexagonal nuts with inner thread
hexagonal nut with inner thread, for Ø 27 mm, ½", PE
Přejít na:
Price
87,70 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
HDPE, white.
  • Good chemical resistance
  • Good temperature resistance
Hexagonal nut with inner thread.
Specifikace
  • Popis:
    hexagonal nut with inner thread, for Ø 27 mm
  • Materiál:
    PE
  • Závit:
    ½"