Microscope slides, Menzel Gläser, SuperFrost®
Adhesive slide, SuperFrost®, Zelená
631-0709
: Epredia
These slides come with a light opaque coating in standard colours.
- Ground 90° edges
- Label areas for easy identification
- Resistant to solvents, and to scratching or sticking when stacked together
Each colour codes different specimens at a glance according to urgency, processing or staining technique.
- Barva:Zelená
- Popis:Adhesive slide, SuperFrost®