Microscope slides, Menzel Gläser, SuperFrost®
Adhesive slide, SuperFrost®, Žlutá
Catalog # 631-0708
Supplier: Epredia
Jump to:
Product Details & Documents
These slides come with a light opaque coating in standard colours.
- Ground 90° edges
- Label areas for easy identification
- Resistant to solvents, and to scratching or sticking when stacked together
Each colour codes different specimens at a glance according to urgency, processing or staining technique.
Documents
Specifications
- Barva:Žlutá
- Popis:Adhesive slide, SuperFrost®