Orders LinkContactUsLinkComponent
Flasks, flat bottom with Quickfit® ground joint
Flasks, flat bottom with Quickfit® ground joint
04, 100 ml, 100 mm, 24/29 NS
Katalogové číslo 201-8432
Dodavatel:   SciLabware
Flasks, flat bottom with Quickfit® ground joint
Katalogové číslo 201-8432
Dodavatel:  SciLabware
Flasks, flat bottom with Quickfit® ground joint
Flasks, flat bottom with Quickfit® ground joint
04, 100 ml, 100 mm, 24/29 NS
Přejít na:
Price
2 510,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
PYREX®, borosilicate glass, clear, with short neck.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
Specifikace
  • Velikost objímky:
    24/29 NS
  • Kapacita:
    100 ml
  • Výška:
    100 mm
  • Sekvence:
    04