Joint clips for conical joints, POM, Quickfit®
03, Žlutá, 14/23
Katalogové číslo 201-8566
Dodavatel: SciLabware
Přejít na:
Product Details & Documents
POM, coloured. Polyacetal clips for retaining glassware having Quickfit® conical joints.
- Easy to fit and remove
- Can be used at temperatures up to +80 °C
- Clips are colour coded for easy identification
Specifikace
- Pro NZ:14/23
- Barva:Žlutá
- Sekvence:03