Spherical ground joints, Quickfit®
, , , -, S 29
Katalogové číslo 201-8208
Dodavatel: SciLabware
Přejít na:
Product Details & Documents
Made of PYREX®, borosilicate glass.
- Excellent sealing properties
- Minimal requirement for joint lubrication
- Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Specifikace
- Kulička:-
- Zásuvka:S 29