Spherical ground joints, Quickfit®
, , , -, S 19
Catalog # 201-8207
Supplier: SciLabware
Jump to:
Product Details & Documents
Made of PYREX®, borosilicate glass.
- Excellent sealing properties
- Minimal requirement for joint lubrication
- Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Specifications
- Kulička:-
- Zásuvka:S 19