Rectangular cover glasses
012, 24×60 mm, 0,13 - 0,16 mm
Katalogové číslo HECH40990166
Dodavatel: GLASWARENFABRIK KARL HECHT
Přejít na:
Product Details & Documents
Cover glass D 263® M made of borosilicate glass.
- Free from colour distortion and highly transparent
- Excellent quality, low levels of inclusions, striae, bubbles and streaks
- High spectral transmission, excellent flatness, optimum resistance to chemicals
Certifications: ISO 8255
Specifikace
- D׊:24×60 mm
- Sekvence:012
- Tloušťka:0,13 - 0,16 mm