Orders LinkContactUsLinkComponent
Cell counters
Cell counters
Benchtop counter, 250×76×56 mm, 6
Katalogové číslo HEATHS5360A
Dodavatel:   HEATHROW
Cell counters
Katalogové číslo HEATHS5360A
Dodavatel:  HEATHROW
Cell counters
Cell counters
Benchtop counter, 250×76×56 mm, 6
Přejít na:
Price
11 240,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These laboratory cell counters can measure cells such as basophils, eosinophils, myelocytes, juvenile, etc.
  • Each key records up to 999
  • Alarm at one hundred totals
  • Red and white buttons with grey housing
They can measure the number of cells such as Basophils (BASO), Eosinophils (EOSIN), Myelocytes (MYELO), Juvenile (JUVEN), Stab., Seg., Lymph., Mono., and total.
Specifikace
  • Popis:
    Benchtop counter, 250×76×56 mm
  • Uspořádání:
    6