Connectors
01, Fittings, external thread for pipes and valves, 1/2"
SEMA4366
: SEMADENI
Connectors for easy installation on any plastic containers by means of counter screw.
- Male thread for fittings or hose connection
- Good chemical resistance to acids and solvents
- Závit:1/2"
- Popis:Fittings, external thread for pipes and valves
- Sekvence:01