Micro capillaries, disposable, minicaps
07, Sodium-heparinised minicaps, 25 µl, 30 - 32 mm
Katalogové číslo 612-2649
Dodavatel: HIRSCHMANN
Přejít na:
Product Details & Documents
Disposable capillary pipettes with the end-to-end system. With genuine Schott DURAN® glass. Accuracy ±≤0,5%, CV ±≤1%. With batch number on the package.
- Correct filling of the capillary
- Conformity certified, according to the German 'Eichordnung' certification
- Colour-coded vial cap with capillary dispenser
- Safe packaging for protection during transportation
Packaging: 100 minicaps packed in a glass vial with dispenser cap. 10 vials plus a pipetting device are packed into a outer carton.
Specifikace
- Popis:Sodium-heparinised minicaps, 25 µl
- Délka:30 - 32 mm
- Sekvence:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



