Volumetric Flasks, Class A
15, 2000 ml, 19/26, 0,60 ± ml
Katalogové číslo WITG3.664.200.BL
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
: Tolerance according to DIN EN ISO 1042
Specifikace
- NZ:19/26
- Kapacita:2000 ml
- Sekvence:15
- Tolerance:0,60 ± ml