Volumetric Flasks, Class A
14, 1000 ml, 24/29, 0,40 ± ml
Katalogové číslo WITG3.664.100.BL
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
: Tolerance according to DIN EN ISO 1042
Specifikace
- NZ:24/29
- Kapacita:1000 ml
- Sekvence:14
- Tolerance:0,40 ± ml