Volumetric Flasks, Class A
11, 200 ml, 14/23, 0,15 ± ml
Katalogové číslo WITG3.664.020.BL
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifikace
- NZ:14/23
- Kapacita:200 ml
- Sekvence:11
- Tolerance:0,15 ± ml