Orders LinkContactUsLinkComponent
Volumetric Flasks, Class A
Volumetric Flasks, Class A
1, 5 ml, 10/19, 0,04 ± ml
Catalog # WITG3.664.015.BL
Supplier:   witeg Labortechnik
Volumetric Flasks, Class A
Catalog # WITG3.664.015.BL
Supplier:  witeg Labortechnik
Volumetric Flasks, Class A
Volumetric Flasks, Class A
1, 5 ml, 10/19, 0,04 ± ml
Price
552,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
  • DE-M marked according to the German calibration law
  • Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifications
  • NZ:
    10/19
  • Kapacita:
    5 ml
  • Sekvence:
    1
  • Tolerance:
    0,04 ± ml