Volumetric Flasks, Class A
6, 20 ml, 10/19, 0,04 ± ml
Katalogové číslo WITG3.664.002.BL
Dodavatel: witeg Labortechnik
Přejít na:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifikace
- NZ:10/19
- Kapacita:20 ml
- Sekvence:6
- Tolerance:0,04 ± ml