Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
04, 500 ml, 29/32 NS
Katalogové číslo WITG2.647.500
Dodavatel:   witeg Labortechnik
Separating funnels, Squibb
Katalogové číslo WITG2.647.500
Dodavatel:  witeg Labortechnik
Separating funnels, Squibb
Separating funnels, Squibb
04, 500 ml, 29/32 NS
Přejít na:
Price
2 940,00 Kč
Přihlásit se pro dostupnost
1 * 2 KS
Product Details & Documents
Separating funnels, Squibb, made of borosilicate glass 3.3, with NS stopcock plugs and solid lass key. With PE-stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    500 ml
  • Sekvence:
    04