Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
05, 1000 ml, 29/32 NS
Catalog # WITG2.647.001
Supplier:   witeg Labortechnik
Separating funnels, Squibb
Catalog # WITG2.647.001
Supplier:  witeg Labortechnik
Separating funnels, Squibb
Separating funnels, Squibb
05, 1000 ml, 29/32 NS
Price
3 890,00 Kč
Log in for availability
1 * 2 KS
Product Details & Documents
Separating funnels, Squibb, made of borosilicate glass 3.3, with NS stopcock plugs and solid lass key. With PE-stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    1000 ml
  • Sekvence:
    05