Orders LinkContactUsLinkComponent
Adapters, reduction, Preciso
Adapters, reduction, Preciso
01, , Redukce, 29/32 NS, , 19/26,
Katalogové číslo WITG0.500.005
Dodavatel:   witeg Labortechnik
Adapters, reduction, Preciso
Katalogové číslo WITG0.500.005
Dodavatel:  witeg Labortechnik
Adapters, reduction, Preciso
Adapters, reduction, Preciso
01, , Redukce, 29/32 NS, , 19/26,
Přejít na:
Price
273,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Certifications: According to DIN 12257.
Specifikace
  • Rozměry zábrusu:
    29/32 NS
  • Zásuvka:
    19/26
  • Type:
    Redukce
  • Sekvence:
    01