Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 250-300 ml, 19/26
Catalog # 528-1203
Supplier:   Lenz Laborglas
Dissolved oxygen bottles
Catalog # 528-1203
Supplier:  Lenz Laborglas
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 250-300 ml, 19/26
Price
1 040,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Glass, clear.
  • White inscription label
  • Oblique ground NS stopper
  • Optional stopper clamp is available
For the determination of dissolved oxygen in water according to Winkler. The capacity is engraved on the bottle with an accuracy of ± 0,01 ml. Each bottle is individually adjusted with its stopper (non interchangeable). Each stopper and bottle is identified with the same serial number.
Specifications
  • Kapacita:
    250-300 ml
  • Popis:
    Oxygen bottles acc. to Winkler
  • NZ zátky:
    19/26
Accessories
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • 705-0564
    Clamps NS 14/23 for Dissolved Oxygen Bottles
    Log in for availability
    1 * 1 KS - 218,00 Kč
Product Information
  • Catalog #
    Popis
    Availability
    Price
  • 705-0565
    Clamps NS 19/26 for Dissolved Oxygen Bottles
    Log in for availability
    1 * 1 KS - 245,00 Kč