Pufr redoxní ORP +475 mV, objem 500ml 1 * 500 mL
Catalog # HAMI238227
Supplier: HAMILTON BONADUZ
Frequently Bought Together
Kyselina dusičná 67%, NORMATOM® for trace metal analysis
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Baňky dle Erlenmeyera
Supplier: VWR Collection
Protective laboratory labelling tapes, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
VWR®, Syringe Filters
Supplier: VWR Collection
Kyselina sírová 98% ≥98%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Pasteur pipettes
Supplier: HIRSCHMANN
VWR® NITRILE LIGHT, Rukavice, nitrilové
Supplier: VWR Collection
VWR®, Redox (ORP) Standards
Supplier: VWR Chemicals
Pufr redoxní ORP +271 mV, objem 500ml 1 * 500 mL
Supplier: HAMILTON BONADUZ
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Anion exchange HPLC columns, RCX-30
Supplier: HAMILTON BONADUZ
N,N-Dimethylformamid
Supplier: Lach-ner S.R.O.
VWR®, kultivační médium, připravené k použití, v miskách
Supplier: VWR Chemicals
Kieselguhr filter papers
Supplier: MACHEREY-NAGEL