Jars, with lid
80 mm, 40 mm
Catalog # HECH41050008
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Jars, low form, made of glass, with notch and overlapping cover.
Specifications
- Ø vněj.:80 mm
- Výška:40 mm
Jars, low form, made of glass, with notch and overlapping cover.