Reference electrodes, ScienceLine
Single reference electrode, ScienceLine, with standard taper NS 14,5, Ag/AgCl reference system, B 3520+ Ag/AgCl, 0 - 14, −5...+100 °C, Sklo, KCl 3 mol/l, Platinum, 10×103 mm
Katalogové číslo SCOI1070073
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
Reference electrodes with glass shaft.
Documents
Specifikace
- Ø×D:10×103 mm
- Type:B 3520+ Ag/AgCl
- Popis:Single reference electrode, ScienceLine, with standard taper NS 14,5, Ag/AgCl reference system
- Materiál:Sklo
- Diafragma:Platinum
- Elektrolyt:KCl 3 mol/l
- Tepl.:−5...+100 °C
- pH:0 - 14