Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Pinzety
Forceps
Forceps
Print
Forceps
Curved, pointed, 105 mm
Catalog # 232-2160
Supplier:
RSG
Forceps
Catalog # 232-2160
Supplier:
RSG
Expand image
Return to Top
Forceps
Curved, pointed, 105 mm
Specifications
Add to cart
Jump to:
Specifications
Price
1 190,00 Kč
Log in
for availability
Unit of Measure
1 * 5 KS
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
Documents
Certificates
Search by Lot Number
Datasheet
RSGA611.300_TDS_EN
Specifications
Type:
Curved, pointed
Délka:
105 mm