Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Double spatulas, one curved end

Double spatulas, one curved end

150 mm, 9 mm
Katalogové číslo RSGA038.150
Dodavatel:   RSG
Double spatulas, one curved end
Katalogové číslo RSGA038.150
Dodavatel:  RSG
Double spatulas, one curved end
Double spatulas, one curved end

150 mm, 9 mm

Přejít na:
Price
1 140,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents

Double spatulas made of stainless steel. With one curved spatula end.

Specifikace
  • Délka:
    150 mm
  • Šířka čepele:
    9 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.