Orders LinkContactUsLinkComponent
Wire triangles
Wire triangles
Tube length 40 mm
Catalog # RSGA257.404
Supplier:   RSG
Wire triangles
Catalog # RSGA257.404
Supplier:  RSG
Wire triangles
Wire triangles
Tube length 40 mm
Price
208,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Wire triangles made from stainless steel, with clay pipes.
Specifications
  • Popis:
    Tube length 40 mm