Orders LinkContactUsLinkComponent
Wire triangles
Wire triangles
Tube length 60 mm
Catalog # RSGA257.406
Supplier:   RSG
Wire triangles
Catalog # RSGA257.406
Supplier:  RSG
Wire triangles
Wire triangles
Tube length 60 mm
Price
1 110,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Wire triangles made from stainless steel, with clay pipes.
Specifications
  • Popis:
    Tube length 60 mm