Forceps
Sharp, 100 mm
Katalogové číslo 232-0050
Dodavatel: RSG
Přejít na:
Product Details & Documents
Steel mainspring, lightweight. Special forceps for particularly accurate and delicate work.
- Lightweight
- Available in two designs
- Extremely flexible
Specifikace
- Type:Sharp
- Délka:100 mm